Cultured olfactory bulb neuron. RH 414 and DM-BODIPY(–)-dihydropyridine.
Cultured olfactory bulb neuron stained with RH 414 (T1111) and DM-BODIPY(–)-dihydropyridine (D7443). Left: image at greater than 580 nm; Middle: image at 510–580 nm; Right: ratio of the middle image divided by the left image. Images were acquired with a Leica confocal laser-scanning microscope. The images were contributed by D. Schild, H. Geiling and J. Bischofberger, Physiologisches Institut, Universität Göttingen.
Related Products
Related Images
Cyanobacteria. BODIPY® FL, DAPI and CellTracker™ Green BODIPY®. Go ›
Immunofluorescent Staining using Mouse HuC/HuD Neuronal Protein Monoclonal Antibody, Biotin-XX Conjugate. Go ›
Schistosoma mansoni parasite. Go ›
Immunohistochemistry using GFAP Monoclonal Antibody, Mouse, Alexa Fluor® 594 Conjugate Go ›