Microtubules of bovine pulmonary artery endothelial cells.
Microtubules of bovine pulmonary artery endothelial cells tagged with mouse monoclonal anti–a-tubulin antibody (Cat. No. A11126) and subsequently probed with: Alexa Fluor® 488 Goat Anti–Mouse IgG antibody (Cat. No. A11001, top panel), Alexa Fluor® 546 Goat Anti–Mouse IgG antibody (Cat. No. A11003, middle panel) or Alexa Fluor® 594 Goat Anti–Mouse IgG antibody (Cat. No. A11005, bottom panel). These images were acquired using a fluorescein bandpass optical filter set, a rhodamine bandpass optical filter set, and a Texas Red bandpass optical filter set, respectively.
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Increase in accuracy and precision using the Qubit® Go ›
Performance of the Qubit® dsDNA HS Assay Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›