BPAE detection of oxidative stress using MitoTracker® Green (Cat. No. M7514) and CellROX® Deep Red Reagent (Cat. No. C10422)
BPAE cells were plated in a 96 well plate, treated with 100 µM menadione for 1 hr and then stained with 20 nM MitoTracker® Green, 5 µM Far Red ROS Sensor and Hoechst 33342 for 30 mins in complete medium. Cells were then washed 3X with PBS and imaged on a Thermo Fisher Cellomics ArrayScan® VTI. The control sample has no signal while in menadione treated cells, there is a robust increase in the signal as a result of oxidative stress caused by menadione. Both MitoTracker® Green and Far Red ROS Sensor stained well showing that they work together in a multiplex assay.
Related Products
Related Images
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Oxidative stress analysis in BPAE and Raw cells using CellROX® Deep Red Reagent (Cat. No. C10422) Go ›
BPAE imaging using CellROX® Deep Red Reagent (Cat. No. C10422) Go ›
Live cell imaging with CellLight™ reagents. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›