Real-time PCR kits for the qualitative and quantitative detection of common pathogens that put these patient groups at risk
Immunosuppressed and immunocompromised persons, including solid organ and hematopoietic stem cell and tissue transplant recipients, or persons undergoing cancer treatment or with unsuppressed HIV infection, are at elevated risk of complications from infectious diseases. Such persons require prompt diagnosis and frequent viral load monitoring to guide prophylaxis treatment and care. Academic medical centers and hospital clinical testing laboratories require a comprehensive testing menu of pathogens commonly associated with these patients that deliver accurate and timely results. The TaqPath™ Menu transplant portfolio supports the testing needs of transplant centers with a range of CE-IVD-marked, reliable infectious disease assays specifically designed to address the common pathogens that put these patient groups at risk. Some of these targets include CMV, EBV, BKV, HSV-1/HSV-2, Adenovirus, Aspergillus and several others. 
Rely on a comprehensive portfolio of Applied Biosystems™ TaqPath Menu Transplant and Immunocompromised Kits
 The real-time PCR (qPCR) assays could be used for:
  • Aiding in the diagnosis and management of transplant patients
  • Measuring viral load to inform the need for treatment
  • Monitoring a patient’s response to therapy
Want to use our solutions?  Contact us for details on our limited time offer.
WHO standard–based quantification
  • Precise and fully traceable quantification according to 1st WHO International Standard NIBSC 09/162​
Simple laboratory workflow​
  • Easily combinable with other PCR kits from the TaqPath Menu in one workflow​
  • Compatible with a wide range of real-time PCR devices
CE-IVD marked kit
Easy-to-use concept
  • Single tube, ready-to-use Master Mix contains all components for PCR amplification
  • No additional pipetting of PCR reagents is necessary
Contamination reduction
  • Master Mix contains uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
 Download flyer
WHO standard–based quantification
  • Precise and fully traceable quantification according to WHO 1st International Standard NIBSC 09/260
  • Enables reliable viral load monitoring​
Simple laboratory workflow​
  • Easily combinable with other PCR kits from the TaqPath Menu in one workflow​
Compatible with a wide range of real-time PCR devices
CE-IVD marked kit
Easy-to-use concept
  • Single tube, ready-to-use Master Mix contains all components for PCR amplification
  • No additional pipetting of PCR reagents is necessary
 Download flyer
WHO standard–based quantification
  • Precise and fully traceable quantification according to 1st WHO International Standard NIBSC BK 14/212 and NIBSC JC 14/114)​
  • Enables reliable viral load monitoring
Simple laboratory workflow​
  • Easily combinable with other PCR kits from the TaqPath Menu in one workflow​​
Compatible with a wide range of real-time PCR devices
CE-IVD marked kit
Easy-to-use concept
  • Single tube, ready-to-use Master Mix contains all components for PCR amplification
  • No additional pipetting of PCR reagents is necessary
 Download flyer
WHO standard–based quantification
  • Precise and fully traceable quantification according to 1st World Health Organization (WHO) International Standard NIBSC BKV 14/212
  • Enables reliable viral load monitoring
Simple laboratory workflow
  • Easily combinable with other PCR kits from the TaqPath Menu in one workflow
Easy-to-use concept
  • Single tube, ready-to-use Master Mix contains all components for PCR amplification
  • No additional pipetting of PCR reagents is necessary
Contamination reduction
  • Master Mix contains uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
Compatible with a wide range of real-time PCR devices
CE-IVD marked kit
 Download flyer
Excellent performance
  • High diagnostic sensitivity (100.00%) and diagnostic specificity (97.44%)
  • Quantification of viral load provides additional information for management of VZV infection
Simple laboratory workflow
  • Easily combinable with other PCR kits from the TaqPath Menu in one workflow
Easy-to-use concept
  • Single tube, ready-to-use Master Mix contains all components for PCR amplification
  • No additional pipetting of PCR reagents is necessary
Contamination reduction
  • Master Mix contains uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
Compatible with a wide range of real-time PCR devices
CE-IVD marked kit
 Download flyer
More information from one sample
  • Detects HHV6 and HHV7 in a single reaction
  • Precise quantification of HHV6 and HHV7 in two separate channels
High sensitivity and traceability
  • Sensitivity was established with WHO-certified strain
Simple laboratory workflow
  • Easily combinable with other PCR kits from the TaqPath Menu in one workflow
Contamination reduction
  • Master Mix contains uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
Compatible with a wide range of real-time PCR devices
CE-IVD marked kit, compliant with IVDR
 Download flyer
High performance
  • Dual targeting prevents a detection failure caused by the occurrence of mutations
  • Unique possibility to quantify viral load
Simple laboratory workflow
  • Easily combinable with other PCR kits from the TaqPath Menu in one workflow
Easy-to-use concept
  • Single tube, ready-to-use Master Mix contains all components for PCR amplification
  • No additional pipetting of PCR reagents is necessary
Contamination reduction
  • Master Mix contains uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
Compatible with a wide range of real-time PCR devices
CE-IVD marked kit, compliant with IVDR
 Download flyer
WHO standard–based quantification
  • Precise and fully traceable quantification according to WHO International Standard NIBSC 12/208
  • Enables reliable viral load monitoring
Simple laboratory workflow
  • Easily combinable with other PCR kits from the TaqPath Menu in one workflow
Easy-to-use concept
  • Single tube, ready-to-use Master Mix contains all components for PCR amplification
  • No additional pipetting of PCR reagents is necessary
Contamination reduction
  • Master Mix contains uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
Compatible with a wide range of real-time PCR devices
CE-IVD marked kit
 Download flyer
WHO standard–based quantification
  • Precise and fully traceable quantification according to 1st WHO International Standard NIBSC JCV 14/114
Simple laboratory workflow
  • Easily combinable with other PCR kits from the TaqPath Menu in one workflow
Easy-to-use concept
  • Single tube, ready-to-use Master Mix contains all components for PCR amplification
  • No additional pipetting of PCR reagents is necessary
Contamination reduction
  • Master Mix contains uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
Compatible with a wide range of real-time PCR devices
CE-IVD marked kit
 Download flyer
High performance
  • Secured by targeting the multicopy sequence of the intergenic spacer ITS2/28S rDNA
  • Enables specific antifungal therapy of resistant aspergillosis
Simple laboratory workflow
  • Easily combinable with other PCR kits from the TaqPath Menu in one workflow
Easy-to-use concept
  • Single tube, ready-to-use Master Mix contains all components for PCR amplification
  • No additional pipetting of PCR reagents is necessary
Contamination reduction
  • Master Mix contains uracil-DNA glycosylase (UNG) and dUTPs reducing carryover contamination
Compatible with a wide range of real-time PCR devices
CE-IVD marked kit
 Download flyer
Ordering information
For In Vitro Diagnostic Use.
Intended use and regulatory statements of products vary. For specific intended use and regulatory statements, please refer to the Instructions for Use (IFU).
Product availability by country varies. For product availability in your country, check the associated product webpages. TaqPath™ Menu | GeneProof™ assays are manufactured by GeneProof™ and distributed by Thermo Fisher Scientific.
© 2023 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
Style Sheet for Global Design System