Marine viruses, bacteria, and a diatom.
An environmental sample containing marine viruses (smallest dots), bacteria (larger, brighter dots), and a diatom (long thin cell with prominent nucleus) were stained with SYBR® Green I Nucleic Acid Stain (Cat. No. S7563, S7567, S7585). Image contributed by Jed Fuhrman, University of Southern California.
Related Products
Related Images
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›