Adherent CRE BAG 2 cells. Alexa Fluor 594 hydrazide and Influx pinocytic cell-loading reagent.
Adherent CRE BAG 2 cells passively loaded with the membrane-impermeant polar tracer Alexa Fluor 594 hydrazide (A10438, A10442). The image on the left illustrates the relatively uniform, cytoplasmic labeling one can obtain with the Influx pinocytic cell-loading reagent (I14402), compared to the punctate labeling that results from pinocytic uptake in normal growth medium (right panel). Both images were acquired using a bandpass filter set appropriate for rhodamine dyes.
Related Products
Related Images
Live CRE BAG 2 cells. Go ›
lacZ-negative (NIH 3T3) and lacZ-positive (CRE BAG 2) cells. Go ›
Dual-emission ratiometric measurement of lysosomal pH using LysoSensor™ Yellow/Blue dextran. Go ›
Rhizoid cells from the marine alga Fucus serratus were pressure-microinjected with 10,000 MW Calcium Green™-1 dextran and 10,000 MW Texas Red® dextran. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›