Bovine pulmonary artery endothelial cells.
The mitochondria of bovine pulmonary artery endothelial cells stained with MitoTracker® Red CM-H2XRos (Cat. No. M7513). The cells were subsequently fixed, permeabilized and treated with RNase. Then the nuclei were stained with SYTOX® Green nucleic acid stain (Cat. No. S7020). The multiple-exposure photomicrograph was acquired using a fluorescence microscope equipped with bandpass filter sets appropriate for fluorescein and Texas Red® dyes.
Related Products
Related Images
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›