Follicle of the giant silkmoth Hyalophora cecropia.
The "delta" region of a developing follicle of the giant silkmoth Hyalophora cecropia stained with an antibody against the largest subunit of the Drosophila RNA polymerase II (RNAp II). The primary antibody was visualized using a biotinylated secondary antibody followed by Texas Red® dye–conjugated streptavidin (Cat. No. S872). The distribution of the RNAp II appears violet because the Texas Red® staining colocalizes with the blue autofluorescence of the yolk granules. Image contributed by Ivo Sauman, Wesleyan University.
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›