Freshwater snail embryo. SYTO Blue fluorescent nucleic acid stain, a component of the SYTO Blue Fluorescent Stain Sampler Kit.
The developing embryo of a freshwater snail stained with SYTO 40 blue-fluorescent nucleic acid stain (S11351), a component of the SYTO Blue Fluorescent Stain Sampler Kit (S11350). A series of z-plane images was acquired with a wide-field optical sectioning confocal microscope. A three-dimensional volume rendering was generated from the deconvolved image series. Image contributed by Bruce Roth and Paul Millard, Molecular Probes, Inc., and Hillary MacDonald, Applied Precision, Inc.
Related Products
Related Images
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›