HeLa cell nuclei. Texas Red-X streptavidin, biotin-XX goat anti–mouse IgG antibody, ELF 97 Cytological Labeling Kit and Hoechst 33258.
HeLa cell nuclei incubated with a mouse monoclonal antibody directed against nuclear-localized antigens (a gift from Benjamin Blencowe and Phil Sharp, Massachusetts Institute of Technology) in conjunction with Texas Red-X streptavidin (S6370) and with a human anti-nuclear antibody in conjunction with biotin-XX goat anti–human IgG antibody. The reagents in the ELF 97 Cytological Labeling Kit (E6603) were then used to detect the biotinylated secondary antibody. Cells were counterstained with Hoechst 33258 (H1398, H3569, H21491). Multiple exposures were taken using filters appropriate for the Texas Red dye, fluorescein and DAPI.
Related Products
Related Images
Prostate carcinoma. Go ›
Transformed mouse fibroblasts (CRE BAG 2 cells). Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›