Fixed and permeabilized HeLa cells stained with Alexa Fluor® 350 phalloidin and Mouse anti ATP Synthase Subunit IF1 Monoclonal Antibody to label actin filaments and the mitochondria, respectively.
Fixed and permeabilized HeLa cells stained with Alexa Fluor® 350 phalloidin (Cat. no. A22281) and Mouse anti ATP Synthase Subunit IF1 Monoclonal Antibody (Cat. no. A21355) to label actin filaments and the mitochondria, respectively. An anti–cdc6 peptide antibody (Cat. no. A21286) was used to label the nucleus. The anti–OxPhos Complex V inhibitor protein antibody was labeled with the Zenon® Alexa Fluor® 488 Mouse IgG1 Labeling Kit (Cat. no. Z25002), and the anti–cdc6 peptide antibody was labeled with the Zenon® Alexa Fluor® 568 Mouse IgG1 Labeling Kit (Cat. no. Z25006).
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Fixed and permeabilized bovine pulmonary artery endothelial cells stained with Mouse anti-ATP Synthase Subunit IF1 Monoclonal Antibody. Go ›