Three-color staining of HeLa cells using fluorescent Qdot® nanocrystal conjugates.
Three-color staining of HeLa cells using fluorescent Qdot® nanocrystal conjugates. The intracellular structures in fixed HeLa cells were visualized using a red-fluorescent Qdot® 655 F(ab')2 goat anti-mouse IgG (Q11021MP, Q11022MP) (nuclei), a yellow-fluorescent Qdot® 585 F(ab')2 goat anti-rabbit IgG (Q11411MP) (Golgi), and a green-fluorescent Qdot® 525 streptavidin conjugate (Q10141MP) (microtubules).
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Human tonsil section visualized with fluorescent Qdot® nanocrystal conjugates. Go ›
Porcine primary skeletal muscle cell transduced with Organelle Lights fluorescent proteins. Go ›