Immunocytochemistry analysis using ABfinity™ Recombinant Rabbit anti Alpha-Synuclein Monoclonal Antibody.
Immunocytochemistry analysis of HeLa cells stained with Alpha-Synuclein Rabbit Recombinant Oligoclonal Antibody using (A) Alexa Fluor® 488 Goat Anti-Rabbit was used as secondary (green). (B) DAPI was used to stain the nucleus (blue) and (C) Alexa Fluor® 594 phalloidin was used to stain actin (red). (D) Composite image of cells showing cytoplasmic and nuclear localization of Alpha-Synuclein. (Cat. No. 701085)
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
Increase in accuracy and precision using the Qubit® Go ›
Performance of the Qubit® dsDNA HS Assay Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›
Immunocytochemistry analysis using Nucleostemin ABfinity™ Recombinant Rabbit Oligoclonal Antibody. Go ›
Immunocytochemistry analysis using Connexin 40 (C-term) ABfinity™ Recombinant Rabbit Oligoclonal Antibody. Go ›