">
Less background DNA for low-level microbial detection
Microbial testing is a central pillar of biopharmaceutical quality control (QC) testing programs. With the ability to quickly and reproducibly detect low-level microbial DNA targets, including bacterial, fungal, mammalian, viral, and antibiotic resistance markers, Applied Biosystems TaqPath BactoPure Microbial Detection Master Mix is ideal for biopharma and biotech workflows. This General Purpose Reagent, labeled ‘For Laboratory Use’, is manufactured in an ISO 13485-certified and FDA-registered facility that adheres to the principles of current good manufacturing practices (cGMP).
TaqPath BactoPure Microbial Detection Master Mix, available with and without ROX, is optimized for multiplexing up to four targets per reaction, even in the presence of PCR inhibitors. The wide dynamic range allows the same assay to be used across many workflow steps where tolerances may be different. 
Try TaqPath BactoPure Microbial Detection Master Mix
TaqPath BactoPure master mix can be widely used across all the biologic modalities, including monoclonal antibodies (mAbs), vaccines, cell and gene therapies, large molecule injectables, and other IV drug substances, as well as in drug product development and manufacturing steps, supporting microbe-free production of drug products.
Empower your drug and biologics development and manufacturing units with TaqPath BactoPure master mix to help produce microbe-free drugs and therapeutics with confidence.
Selected resources
">