Superior Leak Tight Performance
The Clicktainer Vials are proven leak tight in accordance with BS EN 14254, validated as 95kPa compliant and rigorously tested for suitability in pneumatic tubes.
Select the product code of interest and it will appear on the top of the ordering page.
*The term “leakproof” applies to Thermo Scientific Samco Clicktainer products, 90mL/53 mm and 120mL/53 mm, that meet the following criteria: a) a vacuum test that draws 95kPa of pressure within a sealed chamber with no leakage; b) leak testing in accordance with BS EN 14254, which involves filling the container with a dye, placing for 2 minutes on a roller mixer followed by 1 hour inverted in a water bath with no leakage. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Samco Clicktainer Vials under conditions of their planned applications. Thermo Scientific Samco Clicktainer Vials are leakproof at ambient temperature and pressure when used with their Clicktainer closures.