Targeted panels
Targets: 2 to 9
Example: SARS-COV-2, Flu A, Flu B, RSV
For Research Use Only. Not for use in diagnostic procedures.
Including bacteria, viruses, and fungi, across multiple sample types
Targets: 2 to 9
Example: SARS-COV-2, Flu A, Flu B, RSV
Targets: 10 to 30
Example: Viral and bacterial targets
For Research Use Only. Not for use in diagnostic procedures.
Learn more about PCR testing formats like single tube and plates ›
Find out more about PCR molecular testing research products. Expand your molecular testing research menu using the PCR instrument you have.
Real-time PCR products with high analytical sensitivity are needed to support laboratory research to study outbreaks and determine disease etiology where pathogens are similar.
Applied Biosystems real-time PCR respiratory research products are used to analyze a range of respiratory pathogens including viruses, bacteria, and fungi, with high analytical sensitivity.
These solutions are efficient, generate informative results, and solve many of the challenges associated with the use of immunoassay-based, culture-based, and other molecular research methods.
Applied Biosystems TrueMark real-time PCR solutions are part of a sample-to-results workflow. Benefits include the ability to:
Flyer:TrueMark SARS-CoV-2, Flu A, Flu B, RSV ›
For Research Use Only. Not for use in diagnostic procedures.
Applied Biosystems TrueMark SARS-CoV-2, Flu A, Flu B, RSV Select Panel
Format: Single-tube multiplex
Targets
Viral | SARS-COV-2 |
Influenza A | |
Influenza B | |
Respiratory syncytial virus (RSV) |
Applied Biosystems TrueMark Respiratory I Plus Panel
Format: Pre-spotted 96-well plates
Targets
Viral | TrueMark Respiratory I Plus Panel | |
Adenovirus (AV) | Influenza B | |
Enterovirus (EV) | Respiratory syncytial virus A/B (RSV) | |
Human Metapneumovirus (hMPV) | Rhinovirus (RV) | |
Influenza A (Pan) | SARS-CoV-2 |
Kit size | 120 samples/10 pack |
Throughput | 12 samples/plate |
Control(s) | RNase P internal control and available positive control |
Sample type(s) | Nasopharyngeal swab |
Turnaround time | ~4 hours |
Instrument compatibility | •QuantStudio 5 Real-time PCR System •QuantStudio 7 Pro Real-time PCR System •Other Applied Biosystems real-time PCR instruments on request |
Software | Design & Analysis Software |
Catalog number | •TrueMark Respiratory I Plus Panel A56284C •TrueMark Custom Panel Request a quote or consultation |
Applied Biosystems TrueMark Respiratory Panel 2.0, TaqMan Array Card
Format: TaqMan Array Card
Targets
Bacteria | Bordetella (Pan) | Haemophilus influenzae | Moraxella catarrhalis | Streptococcus pneumoniae |
Bordetella pertussis | Klebsiella pneumoniae complex | Mycoplasma pneumoniae | ||
Chlamydia pneumoniae | Legionella pneumophila | Staphylococcus aureus | ||
Fungal | Pneumocystis jirovecii | |||
Viral | Adenovirus | Herpesvirus 6 (HHV6) | Influenza B | Parechovirus |
Bocavirus | HHV3 (varicella zoster virus) | Measles virus | Respiratory Syncytial Virus A (RSV A) | |
Coronavirus 229E | HHV4 (Epstein-Barr virus) | MERS-CoV | Respiratory Syncytial Virus B (RSV B) | |
Coronavirus HKU1 | HHV5 (cytomegalovirus) | Mumps virus | Rhinovirus | |
Coronavirus NL63 | Human Metapneumovirus (hMPV) | Parainfluenza virus 1 | SARS-CoV (SARS) | |
Coronavirus OC43 | Influenza A (Pan) | Parainfluenza virus 2 | SARS-CoV-2 | |
Enterovirus D68 | Influenza A/H1-2009 | Parainfluenza virus 3 | ||
Enterovirus Pan | Influenza A/H3 | Parainfluenza virus 4 |
Kit size | 80 samples/10 pack |
Throughput | 8 samples/card |
Control(s) | B. atrophaeus, 18S, Xeno, RNase P, and available positive control |
Sample type(s) | Nasopharyngeal swab, nasopharyngeal aspirate, and bronchoalveolar lavage (BAL) |
Turnaround time | ~5 hours total |
Instrument compatibility | •QuantStudio 7 Real-Time PCR System •QuantStudio 12K Flex Real-Time PCR System |
Software | Relative Quantification Application |
Catalog number |
Applied Biosystems TrueMark Respiratory Panel 2.0, OpenArray Plate
Format: OpenArray Plate
Targets
Bacteria | Bordetella (Pan) | Haemophilus influenzae | Legionella pneumophila | Staphylococcus aureus |
Bordetella pertussis | Klebsiella pneumoniae complex | Mycoplasma pneumoniae | Streptococcus pneumoniae | |
Chlamydia pneumoniae | ||||
Viral | Adenovirus | Herpesvirus 6 (HHV6) | Influenza A/H3 | Respiratory Syncytial Virus A (RSV A) |
Coronavirus 229E | HHV4 (Epstein-Barr virus) | Influenza B | Respiratory Syncytial Virus B (RSV B) | |
Coronavirus HKU1 | HHV5 (cytomegalovirus) | Parainfluenza virus 1 | Rhinovirus | |
Coronavirus NL63 | Human Metapneumovirus (hMPV) | Parainfluenza virus 2 | SARS-CoV-2 | |
Coronavirus OC43 | Influenza A (Pan) | Parainfluenza virus 3 | ||
Enterovirus D68 | Influenza A/H1-2009 | Parainfluenza virus 4 |
Kit size | 480 samples/10 pack |
Throughput | 24 samples/plate |
Control(s) | B. atrophaeus, Xeno, RNase P, and available positive control |
Sample type(s) | Bronchoalveolar lavage, nasopharyngeal swab, or nasopharyngeal aspirate samples |
Turnaround time | ~5 hours |
Instrument compatibility | QuantStudio 12K Flex Real-time PCR System |
Software | Relative Quantification Application |
Catalog number |
Whether you want to modify from an existing design or start from scratch, we're here to walk you through the process.
Ask about our curated, flexible content panels
Custom TrueMark Respiratory II Panel (A55017)
Request a consultation or place an order › Custom TrueMark Respiratory III Panel (A55017)
|
Don't see what you're looking for? We offer customization across all formats so you can create a tailored solution that meets your laboratory's throughput and business needs. |
|
|
|
|
|
|
Format | Single tube assay | Duplex (target and internal control) TaqMan assays, pre-spotted | Singleplex TaqMan assays, pre-spotted | ||
Number of assays | 1 to 5 | 4 and 8 | 12 and 24 | 24 or 48 | 18, 37, or 56 |
Number of samples | 1–384 | 12–24 | 16–32 | 1–8 | 12–48 |
Reaction volume | 10 μL, 20 μL | 10 μL, 20 μL | 10 μL | 1 μL | 33 nL |
Instrument compatibility | Applied Biosystems 7500 Real-time PCR System
Any QuantStudio instrument | QuantStudio 5 Real-Time PCR System | QuantStudio 12K Flex Real-Time PCR System | ||
QuantStudio 7 Pro Real-Time PCR System |
| ||||
Other instruments available on request |
| ||||
Features and benefits | Flexibility in sample throughput; load wells only as needed
| Panels with eight targets offer an intermediate approach to testing | Fast, easy setup
| Lowest cost per pathogen for broad pathogen detection studies |
Maximize your nucleic acid yield for greater assay sensitivity from a range of biological specimens for infectious disease applications with the MagMAX Prime Viral/Pathogen NA Isolation Kit.
For Research Use Only. Not for use in diagnostic procedures.