Immunocytochemistry using Alpha-Tubulin Monoclonal Antibody, Mouse
Binucleate bovine pulmonary artery endothelial cell. Anti–α-tubulin, biotin-XX conjugate; Alexa Fluor® 568 streptavidin and nuclear yellow. A binucleate bovine pulmonary artery endothelial cell labeled with the biotin-XX conjugate of anti–α-tubulin antibody (Cat. no. A21371) and Alexa Fluor® 568 streptavidin (Cat. no. S11226), then counterstained with nuclear yellow (Cat. no. N21485).
Related Products
Related Images
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›