Mouse cerebellum section. NeuroTrace® 530/615 red-fluorescent Nissl stain and nuclear yellow.
Neurons in a mouse cerebellum section labeled with NeuroTrace® 530/615 red-fluorescent Nissl stain (Cat. no. N21482). The Nissl substance, ribosomal RNA associated with the rough endoplasmic reticulum, is specific to neuronal cells. Other cells in the sample are identified with the contrasting nuclear counterstain nuclear yellow (Cat. no. N21485). Smaller neurons appear to be labeled primarily with the nuclear yellow stain. This image is a composite of two micrographs acquired using a DAPI longpass filter set and a filter set appropriate for tetramethylrhodamine.
Related Products
Related Images
Cyanobacteria. BODIPY® FL, DAPI and CellTracker™ Green BODIPY®. Go ›
Immunofluorescent Staining using Mouse HuC/HuD Neuronal Protein Monoclonal Antibody, Biotin-XX Conjugate. Go ›
Developing Drosophila embryo Go ›
Cytoskeleton of a mixed population of granule neurons and glial cells Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›