Bovine pulmonary artery endothelial cell. Anti–bovine a-tubulin mouse monoclonal antibody and Alexa Fluor 532 goat anti–mouse IgG antibody.
A bovine pulmonary artery endothelial cell labeled with mouse monoclonal anti–a-tubulin antibody (A11126) in combination with Alexa Fluor 532 goat anti–mouse IgG antibody (A11002) to stain microtubules. The image was acquired using a bandpass filter set (excitation/emission 535 +- 17.5/590 +- 17.5 nm).
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›