Schistosoma mansoni parasite.
DiI LDL (Cat. No. L3482) bound to the surface and internalized in the gut of the parasite Schistosoma mansoni. The distribution of LDL in the parasite is used to study a putative mechanism by which the parasite may avoid host immune recognition. Image contributed by John P. Caulfield, Harvard School of Public Health.