NIH 3T3 cells showing incorporation of BrdU.
Cells were pulsed with BrdU (Cat. No. B23151) for 30 min before fixation. BrdU incorporated into the DNA of proliferating cells was detected with an anti-BrdU antibody (Cat. No. A21300) and green-fluorescent Alexa Fluor® 488 Goat Anti–Mouse IgG1 isotype–specific secondary antibody (Cat. No. A21121); the BrdU staining pattern is co-localized with the nuclear staining of blue-fluorescent DAPI (Cat. No. D1306, D21490). Mitochondria were labeled with anti-COX, Complex IV, subunit I, and red-fluorescent Alexa Fluor® 594 Goat Anti–Mouse IgG2a isotype-specific antibody (Cat. No. A21135). Coverslips were mounted with Prolong® antifade mounting medium (Cat. No. P7481).
Related Products
Related Images
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Endothelial cells. Go ›
BPAE cells fixed and permeabilized using the Image-iT® Fixation/Permeabilization Kit. Go ›
BPAE cells fixed and permeabilized using the Image-iT® Fixation/Permeabilization Kit. Go ›
Phagocytosis in MMM Cells using pHrodo™ Red E. coli BioParticles® Conjugate® (Catalog # P35361) Go ›
Muntjac skin fibroblast labeled with rhodamine phalloidin. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
Increase in accuracy and precision using the Qubit® Go ›
Performance of the Qubit® dsDNA HS Assay Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›