Labeled paint probes hybridized to human metaphase chromosomes
Labeled paint probes hybridized to human metaphase chromosomes. A biotinylated chromosome 5 probe was detected with Alexa Fluor® 594 streptavidin (Cat. No. S11227), and a digoxigenin-labeled chromosome 2 probe detected with mouse anti-digoxigenin in combination with Alexa Fluor® 488 Goat Anti–Mouse IgG antibody (Cat. No. A11001). Image contributed by Joop Wiegant, Leiden University Medical Center, Leiden, The Netherlands.
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Simultaneous detection of three gene targets in a whole-mount Drosophila embryo by fluorescence in situ hybridization. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›