Comparison of reprogramming workflows
using the CytoTune™ –iPS Sendai Kit (a), mRNA (b) and Lentivirus (c). Generation of iPSCs in three to four weeks is accomplished after a single transduction with CytoTune™ kit while mRNA requires 17 sequential days of transduction. Traditional reprogramming methods such as Lentivirus and Retrovirus can require multiple transductions for iPSC generation.