Detection technologies used for RNA/DNA quantification
DNA and RNA quantification, generally referred to as nucleic acid quantification, is commonly performed to determine the average concentration of DNA or RNA in a sample prior to downstream experiments. Sample purity is also an important consideration to accurately calculate the amount of DNA or RNA in a sample. There are two optical technologies commonly used to quantify nucleic acids: UV-Vis measurement and fluorescence measurement. Choosing the right technology for your samples and workflow results in accurate RNA or DNA quantification and can save significant time and money by helping to prevent downstream experimental failures.
Photometry (UV-Vis) | Fluorescence | |
---|---|---|
How is the optical signal generated? | The photometric measurement of nucleic acids is based on the intrinsic absorptivity properties of nucleic acids (DNA and RNA). When an absorption spectrum is measured, nucleic acids absorb light with a characteristic peak at 260 nm. | The fluorometric measurement of nucleic acids is based upon the use of fluorogenic dyes that bind selectively to DNA or RNA. |
Typical RNA/DNA absorbance spectrum. | Fluorescent dyes selectively bind to DNA, RNA or protein. Dyes only emit signal when bound to the target. | |
How is the optical signal measured? |
|
|
|
| |
How is the concentration of nucleic acids calculated? |
|
|
What are the advantages? |
|
|
What are the disadvantages? |
|
|
We offer a range of instruments to perform nucleic acid quantification using either photometry of fluorescence measurements.
Choice instruments for photometric UV-Vis RNA/DNA quantification
Well-known Thermo Scientific instruments for UV-Vis RNA/DNA quantification include the NanoDrop One/OneC Spectrophotometer for convenient, single-sample microvolume analysis, the NanoDrop Eight Spectrophotometer for 8-sample microvolume analysis, and choice of the Multiskan SkyHigh Microplate Spectrophotometer or the Varioskan LUX Multimode Reader for up to 16 microvolume samples as well as 96-384-well plates. The table below provides additional detail on instrument performance.
NanoDrop One/Onec | NanoDrop Eight | Multiskan SkyHigh | Varioskan LUX | |
---|---|---|---|---|
What is the lower detection limit (dsDNA)? |
| 2.0 ng/µL | Depends on sample format and volume used (in the case of microplates)* See details
96 well plates (normal)
| Depends on sample format and volume used (in the case of microplates)* See details
96 well plates (normal)
|
What is the upper detection limit (dsDNA)? |
| 10000 ng/µL | Depends on sample format and volume used (in the case of microplates) See details
96 well plates (normal)
| Depends on sample format and volume used (in the case of microplates) See details
96 well plates (normal)
|
What are the available sample formats? |
| Drop (1 µL) |
|
|
How many samples can be measured at a time? (throughput) | One sample at a time | Up to 8 | Dependent on assay format See details
| Dependent on assay format See details
|
What volume of sample is required to measure? | 1 µL | 1 µL | As little as 2 µL depending on assay format See details
| As little as 2 µL depending on assay format See details
|
How much time it takes to measure one sample? | 8 seconds | < 20 seconds for 8 samples | Around 1 minute for 16 microvolume samples in the µDrop Plate See detailsWhen performed with the user interface touch screen or SkanIt software (fast mode) | Around 1.3 minutes for 16 microvolume samples in the µDrop Plate See detailsWhen performed with the user interface touch screen or SkanIt software (fast mode) |
Does it provide spectral data of the sample? | Yes | Yes | Yes | Yes |
Does it give information on sample purity? | Yes See detailsIt provides full spectral data (A260/A280 and A260/A230 ratios) as well as Acclaro Contaminant Analysis | Yes See detailsIt provides full spectral data (A260/A280 and A260/A230 ratios) as well as Acclaro Contaminant Analysis | Yes See detailsIt provides full spectral data (A260/A280 and A260/A230 ratios) | Yes See detailsIt provides full spectral data (A260/A280 and A260/A230 ratios) |
Does it detect specific contaminants in the sample? | Yes See detailsAcclaro Sample Intelligence technology detects protein, phenol and guanidine salts and it gives true concentration of the nucleic acids | Yes See detailsAcclaro Sample Intelligence technology detects protein, phenol and guanidine salts and it gives true concentration of the nucleic acids | No See detailsSome information is available, based on the purity ratios | No See detailsSome information is available, based on the purity ratios |
Does it distinguish between DNA and RNA in the sample? | Yes See detailsDistinguishes between mammalian DNA and mammalian RNA | Yes See detailsDistinguishes between mammalian DNA and mammalian RNA | No | No |
Does it support oligonucleotide quantification? | Yes See detailsWith embedded applications and custom methods | Yes See detailsWith embedded applications and custom methods | Yes | Yes |
Does it provide pre-configured protocols? | Yes See detailsThere are user interface touch screen and custom-protocols | Yes See detailsThere are user interface touch screen and custom-protocols | Yes See detailsThere are pre-configured protocols in the interface touch screen and SkanIt software Cloud Library, free with SkanIt Software | Yes See detailsThere are pre-configured protocols in the SkanIt software Cloud Library, free with SkanIt Software |
Does it connect to the Thermo Fisher Connect? | Yes | No | Yes | No |
Does it provide 21CFR Part 11 compliance? | Yes, with optional software | Yes, with optional software | Yes See detailswith SkanIt software Drug Discovery Edition | Yes See detailswith SkanIt software Drug Discovery Edition |
Does it offer robotic automation compatibility? | No | No | Yes | Yes |
How is the instrument operated? | Via a touch screen user interface or via the NanoDrop One PC control software | With a PC software (Nanodrop Eight Operating software) | Via a touch screen user interface or the PC software (SkanIt software) | Via a PC software (SkanIt software) |
What is the overall ease of use? | ++++ | ++++ | ++++ | +++ |
What is the basic protocol? | See details
| See details
| See details
| See details
|
What is the instrument price category? | Medium | Medium | Medium | High |
What are the ownership costs? | No recurring costs | No recurring costs | If using µDrop Plate, no recurring costs; otherwise UV-compatible clear plates | If using µDrop Plate, no recurring costs; otherwise UV-compatible clear plates |
* Theoretical limit of detection (LOD), estimated according to IUPAC, based on the precision of the blanks (3*SD), from the precision specifications of Multiskan SkyHigh and Varioskan LUX microplate readers. |
Choice instruments for fluorescence RNA/DNA quantification
High-performing and popular instruments for performing fluorescence RNA or DNA quantification include Invitrogen Qubit 4 Fluorometer for single-sample microvolume fluorescence measurements, Invitrogen Qubit Flex Fluorometer for medium-throughput fluorescence measurements and the Thermo Scientific Varioskan LUX Multimode Microplate Reader for 96-1536-well microplate monochromator-based fluorescence measurements. The table below provides additional detail on instrument performance.
Qubit 4 | Qubit Flex | Varioskan LUX | ||
---|---|---|---|---|
What is the lower concentration limit (dsDNA)? | 0.005 ng/µL* | 0.005 ng/µL* | Depends on the Quant-iT assay kit, standard concentrations and curve fittings used** See details0.005 ng/µL | |
What is the upper detection limit (dsDNA)? | 4000 ng/µL* | 4000 ng/µL* | Depends on the Quant-iT assay kit and assay conditions | |
What are the available sample formats? | Qubit assay tubes See detailsLow-fluorescence, thin-walled polypropylene tubes | Qubit Flex tube strips See detailsLow-fluorescence, thin-walled PCR tube strips | 96-1536 well plates | |
How many samples can be measured at a time? (throughput) | 1 | Up to 8 | Dependent on assay format See details
| |
What volume of sample is required to measure? | 1-20 µL | 1-20 µL | Dependent on assay format, fluorophore that is measured, among others | |
How much time does it take to measure 1 sample? | <3 seconds | ~3 seconds for 8 samples | Around 1.3 minutes for 16 microvolume samples in the µDrop Plate See detailsWhen performed with the user interface touch screen or SkanIt software (fast mode) | |
Does it give information on sample purity or detect contaminants? | No | No | No | |
Does it distinguish between DNA and RNA in the sample? | Yes See detailsUsing fluorescent dyes that are specific for DNA or RNA | Yes See detailsUsing fluorescent dyes that are specific for DNA or RNA | Yes See detailsUsing fluorescent dyes that are specific for DNA or RNA | |
Does it provide a RNA quality and integrity score? | Yes, when used with Qubit RNA IQ Assay Kit | Yes, when used with Qubit RNA IQ Assay Kit | No | |
Does it support oligonucleotide quantification? | Yes See detailsWith ssDNA assay kit | Yes See detailsWith ssDNA assay kit | Yes See detailsWith Quant-iT OliGreen assay kit | |
Does it provide pre-configured protocols? | Yes See detailsThere are pre-configured protocols in the user interface touch screen | Yes See detailsThere are pre-configured protocols in the user interface touch screen | Yes See detailsThere are pre-configured protocols in the SkanIt Cloud Library, free with SkanIt Software | |
Does it connect to the Thermo Fisher Connect? | Yes | Yes | No | |
Does it provide 21CFR Part 11 compliance? | No | Yes, with optional software | Yes See detailswith SkanIt software Drug Discovery Edition | |
Does it offer robotic automation compatibility? | No | No | Yes | |
What is the overall ease of use? | ++++ | ++++ | ++ | |
What is the basic protocol? | See details
| See details
| See details
| |
What is the instrument price category? | Low | Low-Medium | High | |
What are the ownership costs? |
Qubit tubes |
Qubit Flex tube strips |
non-treated Nunc plates | |
* Reported here are the lower and upper quantitation ranges measured with Qubit dsDNA High sensitivity (HS) kit (Q32851) Qubit dsDNA Broad Range (BR) kit (Cat. No. Q32850) respectively and using variable sample volume (1-20 µL). ** Limit of Detection (LOD) using the Quant-iT dsDNA Assay HS Kit (Cat. No. Q33120) as instructed. LOD estimated according to IUPAC, based on the precision of the blanks (3*SD). Results will vary depending on the curve fittings, standard concentrations and used kits. |
UV-Vis and Vis Spectrophotometry
Meet your analytical challenges with our complete line of ultraviolet-visible (UV-Vis) spectrophotometers. Our award-winning designs and user-friendly software help you quantify, assess purity, and more.
Fluorometers and Fluorescence
Use fluorometers to quantify, detect and monitor analytes and their reactions by measuring the intensity of the fluorescent signal from dyes attached to biological molecules as well as naturally fluorescent molecules based on signature excitation (Ex) and emission (Em) wavelengths.
Ordering information
UV-Vis spectrophotometers
Q: What is the difference between UV and Fluorescence DNA quantification? Which DNA quantification technology should I choose?
A: UV and Fluorescence technologies work differently to quantify DNA. UV quantification relies on the intrinsic absorptivity of DNA and RNA molecules, while fluorescence quantification uses dyes that specifically bind to your molecule of choice. With UV technology, quantification isn't as sensitive, but it has a broader dynamic range and also gives data about sample purity--plus it is faster because there is no reagent prep. With fluorescence technology you get higher sensitivity and molecule-specific data, but it has lower dynamic range and reagent prep is required. The technology you need will depend on what features are important for your lab and experimentation.
Learn more
Q: Which is the best DNA quantification instrument? Which is the best RNA quantification instrument?
A: All of the DNA instruments offered by Thermo Fisher Scientific are of excellent quality. To find the right instrument to quantify DNA or RNA in your lab, explore the detailed comparison of UV-Vis spectrophotometers and fluorometers. Things like sensitivity, throughput and budget may be initial considerations when selecting a DNA quantification instrument. You may also consider whether you need target specificity, sample purity information, RNA quality information, or broad dynamic range.
Support
Nucleic Acid Quantification Support Center
Find tips, troubleshooting help, and resources for your nucleic acid quantification workflow.