Accessing technology to determine structural elucidation affordably is increasingly important in the field of toxicology. Being able to add new compounds is a critical component of toxicology screening analyses. But how can compounds be added quickly without impacting results in lower volume forensic toxicology laboratories?
Overview: Some common techniques for performing toxicology screening analyses include automated immunoassays, gas chromatography-mass spectrometry (GC-MS), and high pressure liquid chromatography-diode array detector (HPLC-DAD). However, these techniques are unable to identify all the toxic compounds potentially present in a urine or plasma sample. Using liquid chromatography-mass spectrometry (LC-MS) for toxicology screening provides several benefits, such as an easy sample preparation procedure and fewer limitations based on compound volatility and thermal stability.
Method: A Thermo Scientific™ LXQ Ion Trap™ mass spectrometer is used to identify unknown compounds in human urine and plasma samples. Samples are analyzed using electrospray ionization (ESI), and HPLC separation is performed with a Thermo Scientific™ Accela™ pump using a Thermo Scientific™ Hypersil™ Gold PFP column. An MS/MS spectral library of 275 drugs and toxic compounds is created, and Thermo Scientific™ ToxID™ software is used to identify compounds present in the sample based on MS/MS spectra and retention times.
Table 1. Thirteen-minute LC method
Time (minutes) | %A | %B |
---|---|---|
0 | 95 | 5 |
0.5 | 95 | 5 |
5.5 | 5 | 95 |
8.5 | 5 | 95 |
8.6 | 5 | 95 |
13 | 95 | 5 |
Results: This LC-MS/MS workflow allows the user to quickly and easily add new analytes to the screening method, with new compounds typically able to be added in less than 1 hour. With ToxID software performing automatic data analysis and reporting, the need for manual data interpretation is eliminated, which increases confidence in compound identification. When compared to other screening methods, the LC-MS/MS screening methodology identifies more analytes.
Custom Image: Double click to configure component
Custom Image: Double click to configure component
Related toxin screening products
Obtain clean, highly reproducible sample extracts with lower elution volumes using sample prep consumables such as Thermo Scientific™ SOLA™ Solid Phase Extraction (SPE) Plates. The enhanced performance gives you greater confidence in your analytical results and lowers operating costs—without compromising ease of use or requiring complex method development.
Substantially simplify LC-MS/MS screening workflows for toxicology laboratories with Thermo Scientific™ ToxID™ Automated Screening Software. Designed for labs where a premium is placed on simple, easy-to-use software for novice LC-MS/MS users, the software provides the sensitivity and specificity required by today's toxicologists.
Resources
Video: World Tour of Innovation
A complete range of Thermo Scientific Ion Trap Mass Specs to Tackle Complex Analytical Challenges