Product details
Ideal for demanding applications that require safe handling of products and protection of personnel and the environment.
Herasafe 2030i Biological Safety Cabinets CTS (Cell Therapy Systems) Series consist of the equipment, a documentation package and compliance services. These packages support your GMP needs for cell and gene therapy manufacturing. They help you get up and running faster, stay compliant, support regulatory audits, and stay on schedule.
Product details
Ideal for demanding applications that require safe handling of products and protection of personnel and the environment.
Documentation package
This product is manufactured within an ISO 13485 environment and is provided with comprehensive FAT documentation to facilitate timely validation.
Compliance services
On-site qualification service options include Installation Qualification (IQ) and Operational Qualification (OQ).
Product and documentation package |
Compliance services* | Catalog Number |
North America (NSF-49)
| IOQPCKE89003610 |
EMEA (EN12469)
| IOQPCKE89003488 |
*Please check availability of compliance services with your local Thermo Fisher Scientific sales representative.
Additionally to the standard USB data output the Thermo Scientific Herasafe 2030i Biological Safety Cabinets (BSCs) have embedded capability to provide cloud-based connectivity. This enables customers to check the BSC status in real time and be alerted to important issues via alarms and alerts. Maintenance requirements are also visible, plus the ability to interrogate the events log.
The BSC is a device with heavy user interaction (almost a work bench). The user can see alarms/alerts first hand via the touchscreen interface and respond as appropriate. The remote app* adds a further dimension of visibility for lab managers and facility managers who can analyze historical data, manage service requirements and measure product utilization.
* Availability of connectivity-enabled models depends on region. Please contact your Thermo Fisher Scientific sales representative to learn more. Thermo Fisher Connect cloud services are currently available in the following countries: Austria, Belgium, Bulgaria, Canada, Croatia, Cyprus, Czech Republic, Denmark, Estonia, Finland, France, Germany, Greece, Hungary, Iceland, Ireland, Italy, Latvia, Liechtenstein, Lithuania, Luxembourg, Malta, Netherlands, Norway, Poland, Portugal, Romania, Slovakia, Slovenia, Spain, Sweden, Switzerland, United Kingdom, United States.
Check BSC status remotely
Analyze usage data
Receive reminders
BSC Connectivity Flyer
This flyer explains the different functionalities available with the Thermo Fisher Connect app available for Herasafe and Maxisafe 2030i BSCs. Read about where to find instrument information, energy consumption, the events log and more.
Step 1 | Step 2 | Step 3 | Step 4 | Step 5 | |
Create a Thermo Fisher Connect account | Download the Instrument Connect app, if desired | Add the BSC to your Wi-Fi network | Register your BSC on Connect account | Start monitoring |
On-Demand Webinar:
“How connected biological safety cabinets can support your lab - best practices today and future trends."
This webinar covers three reasons to connect your biological safety cabinet and current as well as future capabilities of cloud-based connectivity in BSCs.
Equipped with "smart" self monitoring safety features, intuitive graphic touchscreen and built in connectivity to preserve important performance data. The Herasafe 2030i is ideally suited to demanding cell culture applications and other sensitive work requiring assured safety and protection.
Note: This tool is best viewed in Chrome, Firefox or Safari Browsers. Learn more ›