Neural somata from mouse brain section. NeuroTrace 500/525 green-fluorescent Nissl stain, DAPI and CellTracker CM-DiI.
Neural somata from a mouse brain section labeled with NeuroTrace 500/525 green-fluorescent Nissl stain (N21480). Nuclei are labeled with DAPI (D1306, D3571, D21490). Myelin and other lipophilic areas are stained with the red-orange fluorescence from CellTracker CM-DiI (C7000, C7001). The image is a composite of three micrographs acquired using filters appropriate for fluorescein, tetramethylrhodamine and DAPI.
Related Products
Related Images
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Cyanobacteria. BODIPY® FL, DAPI and CellTracker™ Green BODIPY®. Go ›
Immunofluorescent Staining using Mouse HuC/HuD Neuronal Protein Monoclonal Antibody, Biotin-XX Conjugate. Go ›