3T3 cells. Calcein, Influx™ cell-loading reagent.
3T3 cells loaded with calcein (Cat. no. C481) using the Influx™ pinocytic cell-loading reagent (Cat. no. I14402). The image was acquired using a fluorescence microscope equipped with a cooled CCD camera and a bandpass filter set appropriate for fluorescein.
Related Products
Related Images
Live CRE BAG 2 cells. Go ›
lacZ-negative (NIH 3T3) and lacZ-positive (CRE BAG 2) cells. Go ›
Dual-emission ratiometric measurement of lysosomal pH using LysoSensor™ Yellow/Blue dextran. Go ›
Rhizoid cells from the marine alga Fucus serratus were pressure-microinjected with 10,000 MW Calcium Green™-1 dextran and 10,000 MW Texas Red® dextran. Go ›