Nuclear deformation of an apoptotic cell visualized with the SYTOX® Green dye.
Nuclear deformation of an apoptotic cell visualized with the SYTOX® Green dye (S7020). Bovine pulmonary artery endothelial cells were treated with camptothecin for 24 hours, stained with the SYTOX® Green nucleic acid stain and photographed using a fluorescence microscope equipped with a bandpass filter set designed for fluorescein-like dyes.
Related Products
Related Images
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›