Two-color confocal image of a human epidermal whole mount.
Two-color confocal image of a human epidermal whole mount. ß1 integrin was visualized with the monoclonal antibody P5D2 labeled with the green-fluorescent Alexa Fluor® 488 dye using the Alexa Fluor® 488 Monoclonal Antibody Labeling Kit (Cat. No. A20181). a6 integrin was labeled using a mouse monoclonal antibody visualized with Alexa Fluor® 594 Goat Anti–Mouse IgG antibody (Cat. No. A11005) and pseudocolored blue. Image contributed by Uffe Birk Jensen, University of Aarhus, Denmark.
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Increase in accuracy and precision using the Qubit® Go ›
Performance of the Qubit® dsDNA HS Assay Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›