Immunocytochemistry image of A21975, peroxisomal multifunctional enzyme 2 (MFE2) monoclonal antibody.
Human HDFn cells were paraformaldehyde fixed (4%, 20 min) and Triton X-100 permeabilized (0.1%, 15 min). The cells were incubated with the antibody at 5 µg/mL overnight at 4°C. Secondary antibody: (red) Alexa Fluor® 594 Goat Anti-Mouse IgG (H+L) at a 1/1000 dilution for 1 hr. 10% goat serum was used as the blocking agent for all blocking steps. Target protein locates mainly in peroxisome.
Related Products
Related Images
Live cell imaging with CellLight™ reagents. Go ›
Bacteriophage T4 replication proteins stained with SYPRO® Orange Protein Gel Stain. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›