ExtractaBrite EI/CI Ion Source
Robust electron impact (EI) and chemical ionization (CI) performance with a source that is fully removable for maintenance, column change or EI to CI swap without breaking vacuum.
Experience breakthrough performance in high-resolution GC-MS analysis to gain the deepest insights into your sample. The Orbitrap Exploris GC 240 mass spectrometer enables you to:
Achieve ultimate performance and accuracy | Power discovery with ease | Benefit from versatility for diverse analytical challenges |
---|---|---|
Get the right answer the first time and the flexibility to adapt to evolving research demands. The Orbitrap Exploris GC 240 mass spectrometer brings breakthrough 240,000 mass resolving power, MS/MS capability, and leading sensitivity to researchers across all applications. Stay at the forefront of discovery and be confident that you have the very highest data quality and comprehensive quantitative information to advance your research. | Focus on results, not instrument setup. Intuitive instrument control and method templates ensure that exceptional data are accessible to all members of your scientific team. Integrated informatics solutions for quantitation and compound discovery facilitate detailed characterization of your most challenging samples to turn data into scientific understanding. The compact Orbitrap Exploris GC 240 mass spectrometer provides definitive gains in resolution, consistent mass accuracy, and robustness to enhance laboratory capability. | Address diverse analytical challenges today and into the future with ultimate GC-MS performance and flexibility. Expand analytical possibilities with the Thermo Scientific ExtractaBrite electron ionization/chemical ionization (EI/CI), direct analysis probes, MS/MS capability, and GC modularity. Switching between techniques, ion sources, probes, and columns is fast and easy. |
Experience the system that addresses evolving requirements for compound identification and quantitation. Go inside the instrument and view key features and capabilities.
In this article, learn how the Orbitrap Exploris GC 240 mass spectrometer delivers the very highest data quality to accelerate scientific discovery and provide an unprecedented depth of analysis. With outstanding real-world performance and analytical flexibility, the system can answer the most challenging analytical questions.
Find out how GC Orbitrap mass spectrometers are different from other MS systems and can provide a fast track to accurate results and make scientific discoveries. Thermo Scientific GC Orbitrap mass spectrometry is a powerful technology for applications ranging from quantitation, screening, sample profiling, and unknown identification. Find out how users from all application areas are taking advantage of GC Orbitrap MS.
For more than 20 years, Orbitrap technology has been refined to provide advanced performance, day-to-day reliability, and a compact footprint. Now both novice and expert high-resolution MS users can efficiently obtain reliable and accurate results to advance scientific understanding.
Robust electron impact (EI) and chemical ionization (CI) performance with a source that is fully removable for maintenance, column change or EI to CI swap without breaking vacuum.
Reduces noise and increases robustness by preventing neutrals from entering the quadrupole mass filter and eliminating effects of local charging
Improves sensitivity with 0.4 Da FWHM precursor isolation widths
Curved linear trap for precise ion injection
Resolution up to 240,000 at m/z 200 and acquisition rates up to 40 Hz
Facilitates higher energy collisional dissociation (HCD) fragmentation
Thermo Fisher Scientific and Owlstone Medical offer a workflow that provides non-invasive breath testing with reliable sampling and comprehensive results.
The Thermo Scientific TriPlus RSH SMART Autosampler can eliminate the sample preparation bottleneck in GC and GC-MS analytical workflows.
Transform small molecule, full-scan and MSn data into known compounds and molecular pathways using Thermo Scientific Compound Discoverer software.
For screening and quantitation, the Orbitrap Exploris GC mass spectrometer simplifies operations and delivers accurate results.
From small- to large-molecule analyses, the Orbitrap Exploris 240 mass spectrometer delivers performance and versatility to drive discovery and identification with quantitative precision and accuracy.
The Thermo Scientific EXTREVA ASE Accelerated Solvent Extractor offers walkaway automation of your sample preparation workflow, from sample extraction to evaporation.
The world needs science to move faster. Now more than ever. Which is why when more scientists have access to the latest scientific instrumentation, we all benefit. That is the sole purpose of our Lab-Forward Trade-In Program. For a limited time, trade in your chromatography and mass spectrometry instruments and save!