Which process mass spectrometer is best for my application?
Thermo Scientific process mass spectrometers are engineered to meet a number of challenging process applications in the petrochemical, iron and steel, and biotechnology industries. Use this comparison chart to see which process mass spectrometer is right for you.
Prima PRO | Prima BT | |
Measurement Technology | High precision scanning magnetic sector mass spectrometer | High precision scanning magnetic sector mass spectrometer |
Cooling | Air conditioner | Fan cooled |
Ambient Temperature Range | 12 °C to 40 °C | Ideally 18 to 22 °C. Permissible range 12 °C to 30 °C |
Rapid Multistream Sampler | 32 or 64 streams | 16 ports |
RMS Maximum Temperature | 120 °C | 80 °C |
Calibration Manifolds | Up to four 6-way manifolds | Single 6-way manifold |
Number of Serial Ports | 5 | 3 |
Inputs & Outputs | 13 digital inputs, 13 digital outputs plus wide range of optional analog & digital I/O | 4 digital inputs, 3 digital outputs |
Warranty | 3 year standard | 1 year standard, option for 3 years |
Service Kit | Standard | Optional |
Resources
Biotechnology and Pharma
Iron/Steel
Refining/Petrochemical