Reliable total and trace sulphur analysers for oil refineries
Online sulphur analysers replace expensive and time consuming laboratory sampling. Get fast responses and wide measurement ranges of trace sulphur and total sulphur in flare gas, liquid and vapor for reliable emissions monitoring.
Clean Fuels
The superior stability and precision of SOLA iQ as sulphur analyser enables refiners to make timely process adjustments to enhance the economic efficiency of desulphurization and fuel blending operations.
- SOLA iQ Liquid
- SOLA iQ Liquid Trace
Flare Gas & Condensable Vapors
The highly accurate SOLA iQ Flare analyser features a dynamic measuring range from 10 ppm to 100% S by volume with fast high-to-low response time, enabling reliable flare stack sulphur emission reporting. While the SOLA iQ Condensable Vapors measure 5ppm to 1% sulphur and trace sulphur.
- SOLA iQ Condensible Vapors (CV)
- SOLA iQ Condensible Vapors (CV) Trace
- SOLA iQ Flare
Multi-Calibration/Multi-Stream
The SOLA iQ analyser enables multiple streams of different sulphur concentrations (i.e., batch processing, inlet/outlet of reactors, etc.) to be measured by a single analyser.
- SOLA iQ Liquid
- SOLA iQ Vapor
- SOLA iQ Condensible Vapors (CV)
- SOLA iQ Flare
- SOLA iQ Trace
Clean Fuels
The superior stability and precision of SOLA iQ as sulphur analyser enables refiners to make timely process adjustments to enhance the economic efficiency of desulphurization and fuel blending operations.
- SOLA iQ Liquid
- SOLA iQ Liquid Trace
Flare Gas & Condensable Vapors
The highly accurate SOLA iQ Flare analyser features a dynamic measuring range from 10 ppm to 100% S by volume with fast high-to-low response time, enabling reliable flare stack sulphur emission reporting. While the SOLA iQ Condensable Vapors measure 5ppm to 1% sulphur and trace sulphur.
- SOLA iQ Condensible Vapors (CV)
- SOLA iQ Condensible Vapors (CV) Trace
- SOLA iQ Flare
Multi-Calibration/Multi-Stream
The SOLA iQ analyser enables multiple streams of different sulphur concentrations (i.e., batch processing, inlet/outlet of reactors, etc.) to be measured by a single analyser.
- SOLA iQ Liquid
- SOLA iQ Vapor
- SOLA iQ Condensible Vapors (CV)
- SOLA iQ Flare
- SOLA iQ Trace
Determine total sulphur and trace sulphur concentration in liquid or gas phase samples to ensure process optimization, maximize profitability, and maintain regulatory compliance. Utilizing unique Pulsed Ultraviolet Fluorescence (PUVF) technology and the proven Pyrolysis system, the SOLA iQ analyser builds on the success of the SOLA II range of total sulphur analyser and measures from 2ppm to 100% total sulphur content, with detection as low as 25ppb.
Perform total sulphur analysis on liquid hydrocarbon fuel process streams with the Thermo Scientific SOLA II Sulphur Online Analyser. This proven field sulphur analyser addresses the need for a low-maintenance total sulphur and trace sulphur analyser that can reliably measure low ppm levels to help customers meet the tightening, Tier 3, regulatory requirements that limit sulphur levels in fuels.
Perform total sulphur analysis on liquid hydrocarbon fuel process streams with the Thermo Scientific SOLA II Sulphur Online Analyser. This proven field sulphur analyser addresses the need for a low-maintenance total sulphur and trace sulphur analyser that can reliably measure low ppm levels to help customers meet the tightening, Tier 3, regulatory requirements that limit sulphur levels in fuels.
Trade-in your SOLA I, SOLA II or other brand sulfur analyser and select a service offer or cash discount.
Download this free eBook to discover how gas analysis using process mass spectrometers and sulphur analysers monitor flare stack emissions to comply with flare operating limits.
Thermo Scientific AutoCONFIG software is the robust interface of the SOLA iQ. Designed to empower the user, the interface is simple to configure eliminating the need for in depth programming.
Product support and spares
Thermo Fisher Scientific Service Solutions are a vital component of your product’s lifecycle. These comprehensive solutions are exclusively created to drive optimal performance of your time-critical instruments.
When you partner with Thermo Fisher Scientific, you gain access to engineers trained within the same factory your instruments were manufactured. We only ship spare parts that were designed specifically for your instrument. Our expert service team is truly an integrated part of your product’s life cycle.