Digital Rock Analysis: Run advanced geological analysis from your rock imagery data
Digital Rock Analysis ( DRA ) is the study of rock samples by use of rock imagery data with the goal of understanding rock properties.
Digital Rock Analysis encompasses a multi-disciplinary approach involving advanced microscopy and physics combined with geology, geochemistry, petrophysics, and petroleum engineering to understand the pore-scale microstructure of reservoir rock. It is used to help E&P operators reduce exploration and production risk while informing decision-making to improve recovery.
Thermo Scientific PerGeos Software is a comprehensive Digital Rock Analysis solution that provides geologists with advanced analytical tools to better evaluate the reservoir quality.
Rock core plug analysis using X-ray computed tomography
Using whole-core X-ray CT and micro-CT plug data: Understand lamination. Analyze core and heterogeneity logs. Measure fracture density and orientation. Characterize the pore space.
Segment pores, clays, and grains. Run accurate grain size and shape analysis. Perform advanced porosity calculation and pore size distribution analysis.
Digital rock analysis with focused ion beam milling
Focused Ion Beam coupled with Scanning Electron Microscopy systems (FIB-SEMs) are invaluable tools that produce high-resolution imagery of nanoporous structures of tight reservoirs and for multi-scale analysis.
Using FIB/-SEM image data: Quantify pores, clays, organic matter, grains. Perform 3D pore-space characterization. Analyze organic versus mineral porosity. Run 3D size and shape analysis on grains.
Scanning electron microscopy for geological thin section analysis
Scanning Electron Microscopy (SEM) systems allow geologists to capture high-quality images of thin sections of rocks.
Using SEM thin sections: Perform petrography workflows. Run grains size and shape analysis, pore-space characterization. Get high-resolution mineralogy information.
Perform multi-scale geological and petrophysical analysis of your core imaging data
From Core to Pore, using imagery from large CT to FIB/SEM systems, PerGeos Software helps you digitally understand and validate observations and properties of your rock data with a high level of confidence.
Ability to interpret images at multiple resolutions from multiple modalities, advanced 2D/3D and 3D/3D registration tools and adaptive thresholding make PerGeos Software a unique digital platform for powerful up-scaling workflows.
This allows geological and petrophysical properties to be numerically obtained with confidence since all scales of the pore space can be analyzed and interpreted to calculate properties such as global porosity or relative permeability.
PerGeos Core Profile is a dedicated workspace that enables co-visualization of whole-core CT, core photography, and petrophysical logs in a single environment. Core Profile also creates virtual logs based on whole-core CT data for further calibration and communication of how changes in rock type impact how petrophysical properties are measured down-hole.
The PerGeos Software Petrophysics extension provides a powerful suite of advanced models for rock property analysis and simulations to obtain properties such as absolute permeability, formation factor, thermal properties, molecular diffusion, and MICP porosity.
PerGeos Software provides advanced capabilities for two-phase flow simulation. Properties such as capillary pressure, relative permeability resistivity index, or residual oil saturation can be obtained using this powerful, industry-validated technology.
The Pore Network Statistics extension provides a workflow to generate and extract a pore network model (PNM) based on 3D image data. It provides insight into pore and throat size distributions as a natural outcome of producing a pore network model.
Dual Energy Computed Tomography (DECT)
DECT technology adds value to whole-core CT analysis workflows, by providing you with actual density information that can be compared to density logs and used for further characterization of rock formations. This enables you to compare whole-core CT images across multiple formations and wells.
PerGeos Core Profile is a dedicated workspace that enables co-visualization of whole-core CT, core photography, and petrophysical logs in a single environment. Core Profile also creates virtual logs based on whole-core CT data for further calibration and communication of how changes in rock type impact how petrophysical properties are measured down-hole.
The PerGeos Software Petrophysics extension provides a powerful suite of advanced models for rock property analysis and simulations to obtain properties such as absolute permeability, formation factor, thermal properties, molecular diffusion, and MICP porosity.
PerGeos Software provides advanced capabilities for two-phase flow simulation. Properties such as capillary pressure, relative permeability resistivity index, or residual oil saturation can be obtained using this powerful, industry-validated technology.
The Pore Network Statistics extension provides a workflow to generate and extract a pore network model (PNM) based on 3D image data. It provides insight into pore and throat size distributions as a natural outcome of producing a pore network model.
DECT technology adds value to whole-core CT analysis workflows, by providing you with actual density information that can be compared to density logs and used for further characterization of rock formations. This enables you to compare whole-core CT images across multiple formations and wells.
Shorten your learning curve and maximize your investment with this introductory training specifically designed for new users of Amira, Avizo and PerGeos Software.
The course consists of a lecture with hands-on sessions. The training material highlights the basic features and functionalities of Amira, Avizo and PerGeos Software.
Advanced training
Maximize your investment and reduce your time-to-results with this advanced training specifically designed for existing users of Amira, Avizo and PerGeos Software.
The course consists of a lecture with hands-on sessions. The training material highlights advanced features and functionalities of Amira, Avizo and PerGeos Software.
Custom development
With over 25 years of experience in 3D and image processing and hundreds of custom projects delivered to organizations small and large, Thermo Fisher Scientific can provide you with a solution tailored to fit your specific needs.
We can customize and expand our software solutions at various levels.