Methanol-fixed MRC-5 cells stained with SYTO® RNASelect™ green-fluorescent cell stain.
Methanol-fixed MRC-5 cells stained with SYTO® RNASelect™ green-fluorescent cell stain (Cat. no. S32703). Nuclei were stained with DAPI (Cat. no. D1306, D3571, D21490); the densely stained areas are nucleoli.
Related Products
Related Images
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›