Bovine pulmonary artery endothelial cells (BPAEC). ER-Tracker Blue-White DPX.
Live bovine pulmonary artery endothelial cells (BPAEC) stained with ER-Tracker Blue-White DPX (E12353), a Dapoxyl derivative. This image was acquired using a DAPI bandpass optical filter.