Immunocytochemistry using Alpha-Tubulin Monoclonal Antibody, Mouse
Anti–bovine α-tubulin, biotin-XX conjugate; Alexa Fluor 647 goat anti–mouse IgG (H+L) conjugate; SYTOX Orange nucleic acid stain. A zebrafish cryosection incubated with the biotin-XX conjugate of mouse monoclonal anti–α-tubulin antibody (Cat. no. A21371). The signal was amplified with TSA Kit #22, which includes HRP–streptavidin and Alexa Fluor 488 tyramide (Cat. no. T20932). The sample was then incubated with the mouse monoclonal FRet 6 antibody and was visualized with Alexa Fluor 647 goat anti–mouse IgG (Cat. no. A21235), which is pseudocolored magenta. Finally, the nuclei were counterstained with SYTOX Orange nucleic acid stain (Cat. no. S11368).
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Endothelial cells. Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Simultaneous detection of three gene targets in a whole-mount Drosophila embryo by fluorescence in situ hybridization. Go ›
Increase in accuracy and precision using the Qubit® Go ›
Performance of the Qubit® dsDNA HS Assay Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›
Nuclear, cytoskeletal, and mitochondrial staining of Muntjac fibroblasts. Go ›