Microtubules of endothelial cells. Anti–a-tubulin antibody, Alexa Fluor 350 goat anti–mouse IgG antibody, Alexa Fluor 594 phalloidin and Alexa Fluor 488 WGA.
Microtubules of fixed bovine pulmonary artery endothelial cells localized with mouse monoclonal anti–a-tubulin antibody (A11126), which was subsequently visualized with Alexa Fluor 350 goat anti–mouse IgG antibody (A11045). Next, the F-actin was labeled with Alexa Fluor 594 phalloidin (A12381). Finally, the cells were incubated with Alexa Fluor 488 wheat germ agglutinin (W11261) to stain components of endosomal pathways. The superimposed and pseudocolored images were acquired sequentially using bandpass filter sets appropriate for DAPI, the Texas Red dye and fluorescein, respectively.
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›