Fibroblast and neonatal cardiomyocyte. BODIPY FL phallacidin and Texas Red goat anti–mouse IgG (H+L) antibody.
Actin labeled with BODIPY FL phallacidin (B607) and vinculin, a cytoskeletal focal adhesion protein, tagged with a monoclonal anti-vinculin antibody that was subsequently probed with Texas Red goat anti–mouse IgG antibody (T862). The large triangular cell is a fibroblast containing green actin stress fibers terminating in red focal adhesions. The neighboring polygonal cell, a rat neonatal cardiomyocyte, contains green striated actin in the myofibrils terminating in the focal adhesions. The close apposition of the two stains results in a yellowish-orange color. Image contributed by Mark B. Snuggs and W. Barry VanWinkle, University of Texas, Houston.
Related Products
Related Images
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›