Applied Biosystems TaqMan Arrays contain TaqMan Gene Expression Assays dried down in three array formats: 96- and 384-well TaqMan Array plates, 384-well TaqMan Array cards (microfluidic cards), and OpenArray plates. These qPCR arrays are ideal for gene expression profiling and verification applications that require the analysis of tens to thousands of targets.
Regardless of which format you choose, there is a broad selection of content options available, including inventoried (preconfigured), made-to-order flexible, custom, and specialty designed arrays.
TaqMan Array content options
Inventoried
We offer a number of preconfigured, off-the-shelf, 96-well plates or 384-well microfluidic cards that feature the most popular TaqMan Gene Expression assays targeting relevant genes for specific biological pathways, processes, or diseases. The advantage of these panels is they ship in 1–2 days and you can order as few as one plate or 2–4 cards.
Use the TaqMan Assay search tool below to search TaqMan Array cards or plates, or browse the complete list of our inventoried TaqMan arrays below ›
Flexible (made-to-order)
If you don’t want to design your panel from scratch, our flexible content TaqMan Arrays are the perfect choice. Start by selecting a panel of commonly studied pathways, diseases, biological processes, or cellular functions (human, mouse, or rat). Each panel is pre-populated with expertly chosen assays and serves as a starting point for modification using our simple online configuration tool. Within the tool, you can easily swap out genes of interest, reposition assays, or change the layout to fit your experimental design. Minimum order quantity is 6 plates or 10 cards.
Custom
Customized cards and plates allow you to configure your panel from scratch choosing from any of our 2.8 million predesigned TaqMan Gene Expression assays. Upload your assay list and our simple-to-use online configuration tool makes it as easy as dragging and dropping your assay. Minimum order quantity is 6 plates or 10 cards.
Configure Custom TaqMan Array plate (96-well 0.1 mL or 0.2 mL) ›
Configure Custom TaqMan Array card (384-well) ›
Configure Custom OpenArray plate ›
Specialty
Need a custom TaqMan Array that includes custom TaqMan Gene Expression assays, TaqMan SNP Genotyping assays, TaqMan Copy Number assays, TaqMan miRNA assays, or a combination of assay types? The Specialty TaqMan plating service provides TaqMan Assays in 96- or 384-well plates, or 384-well microfluidic cards configured to your specifications.
Search for pre-configured TaqMan Array cards and plates
Not sure what format to use?
Flexible, made-to-order TaqMan Arrays
- Curated list of suggested assays preloaded to get you started
- Can modify as needed
Custom TaqMan Arrays
- Start with a blank slate
- Load any predesigned TaqMan Gene Expression Assay
Instrument compatibility table
Product format/ instrument | QuantStudio 3 | QuantStudio 5 | QuantStudio 6 | QuantStudio 7 | QuantStudio 12k Flex | StepOne Plus | 7500 | 7500Fast | Viia7 |
96-well plate, standard | ✓* | ✓** | ✓ | ✓ | ✓ | ✓ | ✓ | ||
96-well plate, Fast | ✓* | ✓** | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | |
384-well plate | ✓** | ✓ | ✓ | ✓ | ✓ | ||||
Microfluidic cards (TaqMan Array card) | ✓ | ✓ | ✓ | ||||||
OpenArray | ✓ |
* QuantStudio 3 comes with either a 96-well standard OR 96-well fast block
** QuantStudio 5 comes with either a 96-well standard, 96-well fast block OR 384-well block
TaqMan Array Plates on Bio-Rad CFX Connect and CFX96 Touch instruments
Did you know that TaqMan Array plates are compatible with Bio-Rad instruments? It’s true.
How to order TaqMan Array plates based on the model and block of your Bio-Rad instrument
BioRad CFX Connect/CFX96 Touch 0.1 -mL blocks are compatible with our Fast 0.1 -mL 96-well TaqMan Array plates, and are available in all three formats: custom, made-to-order flexible, and inventoried panels.
Learn more about running TaqMan Assays on your Bio-Rad instrument ›
View and download a list of TaqMan Array products comparable to Bio-Rad PrimePCR arrays ›
All other BioRad instruments/blocks are compatible with Specialty TaqMan Array plates, 96-well and 384-well. Contact our Specialty Plating team for more information. If you are still unsure of what product to use contact Technical Support.