Quickly connect LC modules, valves, and columns using Thermo Scientific Viper and nanoViper Fingertight Fittings for leak-free fluidic HPLC and UHPLC connections, which minimize dead volume to improve chromatographic results. Viper and nanoViper Fingertight Fittings require no tools for installation, fit narrow connections (e.g., 10-port valves), are reusable, and combine simplicity with high performance.
Viper and nanoViper Fingertight Fittings support pressures up to 1500 bar (dependent on type) and are made of stainless steel (SST) or biocompatible material such as MP35N, PEEK, or PEEK-shielded fused silica.
|
|
|
Description | ||
Viper and nanoViper capillaries in stainless steel, or biocompatible MP35N, PEEK and PEEK-shielded fused silica. | Pre-assembled kits contain all the HPLC connectors and capillaries needed for running standard or advanced workflows with your HPLC and LC-MS systems. | Connectors, plugs, filters, pre-heaters and post-column coolers, which use Viper Fingertight Fitting technology. |