Mouse brain cryosection. NeuroTrace® 500/525 green-fluorescent Nissl stain and DAPI.
A mouse brain cryosection stained with the neuron-selective NeuroTrace® 500/525 green-fluorescent Nissl stain (Cat. no. N21480). The nuclei of the non-neuronal cells appear blue after incubation with the cell-permeant DNA counterstain, DAPI (Cat. no. D1306, D3571, D21490). The image is a composite of two micrographs acquired using filter sets appropriate for fluorescein and DAPI.
Related Products
Related Images
Cyanobacteria. BODIPY® FL, DAPI and CellTracker™ Green BODIPY®. Go ›
Immunofluorescent Staining using Mouse HuC/HuD Neuronal Protein Monoclonal Antibody, Biotin-XX Conjugate. Go ›
Developing Drosophila embryo Go ›
Cytoskeleton of a mixed population of granule neurons and glial cells Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›