Fixed human epidermoid carcinoma cells (A431). Biotinylated epidermal growth factor (EGF) complexed to Texas Red® streptavidin.
Lightly fixed human epidermoid carcinoma cells (A431) stained with biotinylated epidermal growth factor (EGF) complexed to Texas Red® streptavidin (E3480). An identical cell preparation stained in the presence of a 100-fold excess of unlabeled EGF (E3476) showed no fluorescent signal.
Related Products
Related Images
Schistosoma mansoni parasite. Go ›
Immunohistochemistry using GFAP Monoclonal Antibody, Mouse, Alexa Fluor® 594 Conjugate Go ›
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›