Endogenous alkaline phosphatase activity of osteosarcoma cells. ELF® 97 Endogenous Phosphatase Detection Kit and Hoechst 33342.
Endogenous alkaline phosphatase enzyme of osteosarcoma cells localized with the ELF® 97 Endogenous Phosphatase Detection Kit (Cat. no. E6601). Unlike other phosphatase substrates, the unique ELF® 97 phosphatase substrate forms a fluorescent precipitate at the site of enzymatic activity. The blue-fluorescent nucleic acid stain Hoechst 33342 (Cat. no. H1399, H3570, H21492) was used as a counterstain to the green fluorescence of the ELF® 97 alcohol precipitate. The double-exposure image was acquired using a bandpass filter set appropriate for ELF® 97 alcohol and a longpass filter set appropriate for DAPI.
Related Products
Related Images
Prostate carcinoma. Go ›
Transformed mouse fibroblasts (CRE BAG 2 cells). Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›