Single-strand conformation polymorphism (SSCP) in exon 1 of human K-ras.
Sensitive and direct visualization of single-strand conformation polymorphism (SSCP) in exon 1 of human K-ras using SYBR® Gold Nucleic Acid Gel Stain (Cat. No. S11494). Lane 1 contains wild-type DNA and lanes 2–4 contain DNA from various adenocarcinoma samples with mutant alleles. Image contributed by Valerie DeGroff and Chris Weghorst, Ohio State University.
Related Products
Related Images
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›