Human cheek epithelial cells. Alexa Fluor® 350 wheat germ agglutinin and SYTOX® Green nucleic acid stain.
Human cheek epithelial cells labeled with Alexa Fluor® 350 wheat germ agglutinin (Cat. no. W11263) and stained with SYTOX® Green nucleic acid stain (Cat. no. S7020). This multiple-exposure image was acquired using bandpass filter sets appropriate for DAPI and fluorescein.
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›