Mitotic spindles from sea urchin eggs. YOYO®-1 iodide and Texas Red® goat anti–rat IgG (H+L) antibody.
Mitotic spindles isolated from sea urchin eggs that are labeled with YOYO®-1 iodide (Cat. no. Y3601) and a monoclonal anti–ß-tubulin antibody in conjunction with Texas Red® goat anti–rat IgG antibody (Cat. no. T6392). This image was generated by epifluorescence microscopy. Image contributed by John Murray, University of Pennsylvania.
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›