Drosophila ovarian egg chamber.
Day 10 of development of a Drosophila ovarian egg chamber assembly line. The nuclei of follicle and nurse cells were labeled with propidium iodide (Cat. No. P1304MP, P3566, P21493) and visualized by confocal laser-scanning microscopy using excitation by the 568 nm spectral line of an Ar–Kr laser. Image contributed by Sandra Orsulic, University of North Carolina at Chapel Hill.
Related Products
Related Images
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›