Bovine pulmonary artery endothelial cells. SYTOX® Green nucleic acid stain and BODIPY® TR-X phallacidin.
Permeabilized bovine pulmonary artery endothelial cells stained with SYTOX® Green nucleic acid stain (S7020) to label the nuclei and with BODIPY® TR-X phallacidin (B7464) to label the F-actin. The image was acquired by taking sequential exposures through bandpass optical filter sets appropriate for fluorescein and the Texas Red® dye.
Related Products
Related Images
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›